Invitrogen
Manufacturer Code:PA579298
Catalog # PIPA579298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human GALT recombit protein (Position: Q188-A379). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gal-1-P uridylyltransferase; galactose-1-phosphate uridyl transferase; Galactose-1-phosphate uridylyltransferase; Galactose-1-phosphate uridylyltransferase Gal-1-P uridylyltransferase UDP-glucose--hexose-1-phosphate uridylyltransferase GALT; UDP-glucose--hexose-1-phosphate uridylyltransferase
Gene Aliases: AW553376; GALT
UniProt ID: (Human) P07902, (Mouse) Q03249, (Rat) P43424
Entrez Gene ID: (Human) 2592, (Mouse) 14430, (Rat) 298003
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.