Novus Biologicals
Manufacturer Code:NBP154980
Catalog # NBP154980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to GALT(galactose-1-phosphate uridylyltransferase) The peptide sequence was selected from the C terminal of GALT. Peptide sequence LLRSATVRKFMVGYEMLAQAQRDLTPEQAAERLRALPEVHYHLGQKDRET. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.7.12 Gal-1-P uridylyltransferase galactose-1-phosphate uridyl transferase galactose-1-phosphate uridylyltransferase UDP-glucose--hexose-1-phosphate uridylyltransferase; Gal-1-P uridylyltransferase; galactose-1-phosphate uridyl transferase; Galactose-1-phosphate uridylyltransferase; UDP-glucose--hexose-1-phosphate uridylyltransferase
Gene Aliases: GALT
UniProt ID: (Human) P07902
Entrez Gene ID: (Human) 2592
Molecular Function: nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.