Invitrogen
Manufacturer Code:PA577442
Catalog # PIPA577442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide SYYGTVRYGALELC corresponding to amino acid residues 159-172 of mouse Galanin receptor type 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAL3-R; Galanin receptor type 3; GALR-3; GALR3
Gene Aliases: Galnr3; Galr3
UniProt ID: (Mouse) O88853, (Rat) O88626
Entrez Gene ID: (Mouse) 14429, (Rat) 29235
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.