Invitrogen
Manufacturer Code:PA533578
Catalog # PIPA533578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from 1st cytoplasmic domain of human GALR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAL3-R; Galanin receptor type 3; galanin receptor, family member 3; GALR-3; GALR3 Gal receptor-3 Galanin r3 receptor Gal-r3 Galanin receptor 3 Galanin receptor galr3 Galanin receptor subtype 3 Gal3 receptor GAL3-R Galanin 2 receptor homolog Galanin receptor type 3 GALNR3 GALR-3
Gene Aliases: GALNR3; GALR3
UniProt ID: (Human) O60755, (Rat) O88626
Entrez Gene ID: (Human) 8484, (Rat) 29235
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.