Invitrogen
Manufacturer Code:PA533575
Catalog # PIPA533575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from 7th transmembrane domain of human GALR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAL2-R; Galanin receptor type 2; GALR2 Galanin-2 receptor Gal-2 receptor Galanin receptor type 2 GALNR2 GALR-2 Gal receptor 2 GAL subtype 2 receptor Gal-r2 Galanin r2 receptor GAL2-R Galanin receptor 2 Galanin receptor subtype 2
Gene Aliases: GAL2-R; GALNR2; GALR-2; GALR2
UniProt ID: (Human) O43603
Entrez Gene ID: (Human) 8811
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.