Proteintech
Manufacturer Code:264591AP150UL
Catalog # 50-173-5344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GALR2 Fusion Protein Ag24162 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAL2 R galanin receptor 2 Galanin receptor type 2 GALNR2 GALR 2 GALR2; GAL2-R; Galanin receptor type 2; GALR-2
Gene Aliases: GAL2-R; GALNR2; GALR-2; GALR2; mGalR
UniProt ID: (Human) O43603, (Mouse) O88854
Entrez Gene ID: (Human) 8811, (Mouse) 14428
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.