Proteintech
Manufacturer Code:169001AP150UL
Catalog # 50-173-1246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GALNTL2 Fusion Protein Ag10434 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNAc T like protein 2 GALNT15 GALNT7 GALNTL2 PIH5 pp GaNTase like protein 2; GalNAc-T-like protein 2; polypeptide GalNAc transferase 15; Polypeptide GalNAc transferase-like protein 2; polypeptide N-acetylgalactosaminyltransferase 13; Polypeptide N-acetylgalactosaminyltransferase 15; Polypeptide N-acetylgalactosaminyltransferase-like protein 2; pp-GaNTase-like protein 2; pregnancy-induced hypertension syndrome-related protein 5; Protein-UDP acetylgalactosaminyltransferase-like protein 2; UDP-GalNAc transferase T15; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 7; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 2
Gene Aliases: GALNACT15; GALNT13; GALNT15; GALNT7; GALNTL2; PIH5; pp-GalNAc-T15; UNQ770/PRO1564
UniProt ID: (Human) A6NMN1
Entrez Gene ID: (Human) 117248
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.