Invitrogen
Manufacturer Code:PA564163
Catalog # PIPA564163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GALNT7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNAc-T7; GALNAC-T7 GalNAcT7 N-acetylgalactosaminyltransferase 7 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 7 polypeptide GalNAc transferase 7 polypeptide N-acetylgalactosaminyltransferase 7 pp-GaNTase 7 protein-UDP acetylgalactosaminyltransferase 7; N-acetylgalactosaminyltransferase 7; Polypeptide GalNAc transferase 7; pp-GaNTase 7; Protein-UDP acetylgalactosaminyltransferase 7; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 7; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 7 (GalNAc-T7)
Gene Aliases: GALNAC-T7; GalNAcT7; GALNT7
UniProt ID: (Human) Q86SF2
Entrez Gene ID: (Human) 51809
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.