Proteintech
Manufacturer Code:167161AP150UL
Catalog # 50-173-1133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GALNT3 Fusion Protein Ag10140 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNAc T3 GALNT3 HFTC HHS pp GaNTase 3; GalNAc transferase 3; GalNAc-T3; Polypeptide GalNAc transferase 3; polypeptide GalNAc-transferase T3; Polypeptide N-acetylgalactosaminyltransferase 3; pp-GaNTase 3; ppGaNTase-T3; Protein-UDP acetylgalactosaminyltransferase 3; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3)
Gene Aliases: GalNAc-T3; GALNT3; HFTC; HHS
UniProt ID: (Human) Q14435, (Mouse) P70419
Entrez Gene ID: (Human) 2591, (Rat) 366061, (Mouse) 14425
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.