Invitrogen
Manufacturer Code:PA521541
Catalog # PIPA521541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 55 and 369 (Q10471) of GALNT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNAc-T2; Polypeptide GalNAc transferase 2; Polypeptide N-acetylgalactosaminyltransferase 2; Polypeptide N-acetylgalactosaminyltransferase 2 soluble form; pp-GaNTase 2; ppGaNTase-T2; Protein-UDP acetylgalactosaminyltransferase 2; RP5-956O18.1 GalNAc-T2 UDP-GalNAc transferase 2 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2 polypeptide GalNAc transferase 2 polypeptide N-acetylgalactosaminyltransferase 2 pp-GaNTase 2 protein-UDP acetylgalactosaminyltransferase 2; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2 (GalNAc-T2)
Gene Aliases: AI480629; GalNAc-T2; GALNT2
UniProt ID: (Human) Q10471, (Mouse) Q6PB93
Entrez Gene ID: (Human) 2590, (Mouse) 108148, (Rat) 292090
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.