Novus Biologicals
Manufacturer Code:NBP183394
Catalog # NBP183394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SCKPFKWYLENVYPELRVPDHQDIAFGALQQGTNCLDTLGHFADGVVGVYECHNAGGNQEWALTKEKSVKHMDLCLTVVDRAPGSLIKLQGCRENDSRQKWEQIEGNSKLRHVGSNLCLDSRTAKSGGLSVEVCGPALSQQWKFTLNL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNAc-T2; GalNAc-T2EC 2.4.1.41 Polypeptide GalNAc transferase 2 polypeptide N-acetylgalactosaminyltransferase 2 pp-GaNTase 2 Protein-UDP acetylgalactosaminyltransferase 2 UDP-GalNAc transferase 2 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2 UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 2 (GalNAc-T2); Polypeptide GalNAc transferase 2; Polypeptide N-acetylgalactosaminyltransferase 2; Polypeptide N-acetylgalactosaminyltransferase 2 soluble form; pp-GaNTase 2; Protein-UDP acetylgalactosaminyltransferase 2; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2 (GalNAc-T2)
Gene Aliases: GalNAc-T2; GALNT2
UniProt ID: (Human) Q10471
Entrez Gene ID: (Human) 2590
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.