R&D Systems
Manufacturer Code:AF7507SP
Catalog # AF7507SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Polypeptide GalNac Transferase 2/GALNT2 Lys52-Gln571 Accession # Q10471 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNAc-T2; GalNAc-T2 GalNAc-T2EC 2.4.1.41 Polypeptide GalNAc transferase 2 polypeptide N-acetylgalactosaminyltransferase 2 pp-GaNTase 2 Protein-UDP acetylgalactosaminyltransferase 2 UDP-GalNAc transferase 2 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2 UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 2 (GalNAc-T2); Polypeptide GalNAc transferase 2; Polypeptide N-acetylgalactosaminyltransferase 2; Polypeptide N-acetylgalactosaminyltransferase 2 soluble form; pp-GaNTase 2; Protein-UDP acetylgalactosaminyltransferase 2; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2 (GalNAc-T2)
Gene Aliases: GalNAc-T2; GALNT2
UniProt ID: (Human) Q10471
Entrez Gene ID: (Human) 2590
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.