Proteintech
Manufacturer Code:169391AP150UL
Catalog # 50-173-1277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GALNT14 Fusion Protein Ag10470 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNac T10 GalNac T14 GALNT14 GALNT15 pp GaNTase 14; GalNAc-T14; Polypeptide GalNAc transferase 14; Polypeptide N-acetylgalactosaminyltransferase 14; polypeptide N-acetylgalactosaminyltransferase 15; pp-GaNTase 14; Protein-UDP acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14)
Gene Aliases: 0610033M06Rik; GalNac-T10; GalNac-T14; GALNT14; GALNT15; UNQ2434/PRO4994
UniProt ID: (Human) Q96FL9, (Mouse) Q8BVG5
Entrez Gene ID: (Human) 79623, (Rat) 313878, (Mouse) 71685
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.