Novus Biologicals
Manufacturer Code:NBP16900820UL
Catalog # NBP16900820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to Galnt11 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 11 (GalNAc-T11)) The peptide sequence was selected from the N terminal of Galnt11. Peptide sequence LGMIFNERDQELR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.1.41 FLJ21634 GALNAC-T11 MGC71630 Polypeptide GalNAc transferase 11 polypeptide N-acetylgalactosaminyltransferase 11 pp-GaNTase 11 Protein-UDP acetylgalactosaminyltransferase 11 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11 UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 11 (GalNAc-T11); GalNAc-T11; Polypeptide GalNAc transferase 11; Polypeptide N-acetylgalactosaminyltransferase 11; pp-GaNTase 11; Protein-UDP acetylgalactosaminyltransferase 11; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 11; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 11 (GalNAc-T11)
Gene Aliases: GALNAC-T11; GALNACT11; GALNT11
UniProt ID: (Human) Q8NCW6
Entrez Gene ID: (Human) 63917
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.