Invitrogen
Manufacturer Code:PA547904
Catalog # PIPA547904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Polypeptide GalNac Transferase 1/GALNT1 Gly41-Phe559 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: GalNAc transferase 1; GalNAc-T1; GALNAC-T1 pp-GaNTase 1 EC 2.4.1.41 GalNAc transferase 1 GALNAC-T1 GalNAc-T1 Polypeptide GalNAc transferase 1 polypeptide N-acetylgalactosaminyltransferase 1 pp-GaNTase 1 Protein-UDP acetylgalactosaminyltransferase 1 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 1 UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 1 (GalNAc-T1); Polypeptide GalNAc transferase 1; Polypeptide N-acetylgalactosaminyltransferase 1; Polypeptide N-acetylgalactosaminyltransferase 1 soluble form; pp-GaNTase 1; Protein-UDP acetylgalactosaminyltransferase 1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 1; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 1 (GalNAc-T1)
Gene Aliases: GALNAC-T1; GALNT1
UniProt ID: (Human) Q10472
Entrez Gene ID: (Human) 2589
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.