OriGene
Manufacturer Code:TA812316
Catalog # 50-168-4516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F8 |
Immunogen | Full length human recombit protein of human GALM produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aldose 1-epimerase; aldose mutarotase; BLOCK25 GLAT HEL-S-63p IBD1; epididymis secretory sperm binding protein Li 63p; galactomutarotase; galactose enzyme activator; Galactose mutarotase; galactose mutarotase (aldose 1-epimerase)
Gene Aliases: BLOCK25; GALM; GLAT; HEL-S-63p; IBD1
UniProt ID: (Human) Q96C23
Entrez Gene ID: (Human) 130589
Molecular Function: epimerase/racemase isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.