Novus Biologicals
Manufacturer Code:NBP216592
Catalog # NBP216592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human GALM. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldose 1-epimerase; aldose 1-epimerase BLOCK 25 EC 5.1.3.3 galactomutarotase Galactose mutarotase galactose mutarotase (aldose 1-epimerase) IBD1; aldose mutarotase; epididymis secretory sperm binding protein Li 63p; galactomutarotase; galactose enzyme activator; Galactose mutarotase; galactose mutarotase (aldose 1-epimerase)
Gene Aliases: BLOCK25; GALM; GLAT; HEL-S-63p; IBD1
UniProt ID: (Human) Q96C23
Entrez Gene ID: (Human) 130589
Molecular Function: epimerase/racemase isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.