Proteintech
Manufacturer Code:151641AP150UL
Catalog # 50-173-0033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GALK2 Fusion Protein Ag7278 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Galactokinase 2; galactokinase 2 GALK2 GalNAc kinase GK2 N acetylgalactosamine kinase; GalNAc kinase; N-acetylgalactosamine kinase
Gene Aliases: 2810017M24Rik; AI327030; GALK2; GK2
UniProt ID: (Human) Q01415, (Mouse) Q68FH4, (Rat) Q5XIG6
Entrez Gene ID: (Human) 2585, (Mouse) 69976, (Rat) 296117
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.