Invitrogen
Manufacturer Code:PA513844
Catalog # PIPA513844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 345-375 amino acids from the C-terminal region of human GALK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 19; Galactokinase; Galactokinase Galactose kinase GALK; Galactose kinase
Gene Aliases: AA409894; GALK; GALK1; Gk; GK1; Glk; Glk1; HEL-S-19
UniProt ID: (Human) P51570, (Mouse) Q9R0N0
Entrez Gene ID: (Human) 2584, (Mouse) 14635
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.