Novus Biologicals
Manufacturer Code:NBP299783B
Catalog # NB086261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant GALK1 (P51570 Met1-Leu392). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.6 galactokinase galactokinase 1 Galactose kinase GALK GK1; epididymis secretory protein Li 19; Galactokinase; Galactose kinase
Gene Aliases: GALK; GALK1; GK1; HEL-S-19
UniProt ID: (Human) P51570
Entrez Gene ID: (Human) 2584
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.