Invitrogen
Manufacturer Code:PA579296
Catalog # PIPA579296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human GALE recombit protein (Position: M1-N340). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: galactose-4-epimerase, UDP; galactose-4-epimerase, UDP-; Galactowaldenase; short chain dehydrogenase/reductase family 1E, member 1; UDP galactose-4'-epimerase; UDP-galactosamine 4-epimerase; UDP-galactose 4-epimerase; UDP-GalNAc 4-epimerase; UDP-GlcNAc 4-epimerase; UDP-glucose 4-epimerase; UDP-N-acetylgalactosamine 4-epimerase; UDP-N-acetylglucosamine 4-epimerase
Gene Aliases: 2310002A12Rik; AI323962; GALE; SDR1E1
UniProt ID: (Human) Q14376, (Mouse) Q8R059
Entrez Gene ID: (Human) 2582, (Rat) 114860, (Mouse) 74246
Molecular Function:
dehydratase
epimerase/racemase
isomerase
lyase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.