Invitrogen
Manufacturer Code:MA191587
Catalog # PIMA191587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G152 |
Immunogen | Recombit fragment corresponding to amino acids 1-147 of S. cerevisiae GAL4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: galactose-responsive transcription factor GAL4; Regulatory protein GAL4; YPL248C GAL81
Gene Aliases: GAL4; GAL81; YPL248C
UniProt ID: (Baker's yeast) P04386
Entrez Gene ID: (Baker's yeast) 855828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.