Invitrogen
Manufacturer Code:MA524391
Catalog # PIMA524391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4E4 |
Immunogen | GAL3ST4 (NP&lowbar078913.3 388 a.a. &bsim 486 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-galactose-3-O-sulfotransferase 4; beta-galactose-3-O-sulfotransferase, 4; Gal-beta-1,3-GalNAc 3'-sulfotransferase; Gal3ST-4; GAL3ST-4 beta-galactose-3-O-sulfotransferase 4 beta-galactose-3-O-sulfotransferase 4 gal-beta-13-GalNAc 3&prime-sulfotransferase galbeta1-3GalNAc 3&prime-sulfotransferase; Galactose-3-O-sulfotransferase 4; galbeta1-3GalNAc 3'-sulfotransferase
Gene Aliases: GAL3ST-4; GAL3ST4; PP6968
UniProt ID: (Human) Q96RP7
Entrez Gene ID: (Human) 79690
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.