Invitrogen
Manufacturer Code:PA569637
Catalog # PIPA569637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human GAL3ST4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-galactose-3-O-sulfotransferase 4; beta-galactose-3-O-sulfotransferase, 4; Gal-beta-1,3-GalNAc 3'-sulfotransferase; Gal3ST-4; GAL3ST-4 beta-galactose-3-O-sulfotransferase 4 beta-galactose-3-O-sulfotransferase 4 gal-beta-1 3-GalNAc 3&prime-sulfotransferase galbeta1-3GalNAc 3&prime-sulfotransferase; Galactose-3-O-sulfotransferase 4; galbeta1-3GalNAc 3'-sulfotransferase
Gene Aliases: 1500031A01Rik; GAL3ST-4; GAL3ST4; PP6968; RGD1561105
UniProt ID: (Human) Q96RP7, (Bovine) Q5E9W5
Entrez Gene ID: (Human) 79690, (Rabbit) 100351725, (Bovine) 520552, (Dog) 489843, (Horse) 100067323, (Mouse) 330217, (Rat) 498166, (Guinea pig) 100718399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.