Proteintech
Manufacturer Code:248511AP150UL
Catalog # 50-173-4576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GAL3ST3 Fusion Protein Ag20692 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-galactose-3-O-sulfotransferase 3; Gal-beta-1, 3-GalNAc 3'-sulfotransferase 3; GAL3ST 3 GAL3ST2 GAL3ST3; Gal3ST-3; Gal3ST3; galactose 3'-sulfotransferase; Galactose-3-O-sulfotransferase 3; galbeta1-3GalNAc 3'-sulfotransferase 3
Gene Aliases: GAL3ST-3; GAL3ST2; GAL3ST3
UniProt ID: (Human) Q96A11, (Mouse) P61315
Entrez Gene ID: (Human) 89792, (Mouse) 545276, (Rat) 499302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.