Invitrogen
Manufacturer Code:PA564472
Catalog # PIPA564472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GAL3ST2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-galactose-3-O-sulfotransferase 2; gal 3-O-sulphotransferase; Gal-beta-1, 3-GalNAc 3'-sulfotransferase 2; Gal3ST-2; GAL3ST-2 GP3ST beta-galactose-3-O-sulfotransferase 2 gal 3-O-sulphotransferase gal-beta-1 3-GalNAc 3&prime-sulfotransferase 2 galbeta1-3GalNAc 3&prime-sulfotransferase 2 glycoprotein beta-Gal 3&prime-sulfotransferase 2; Galactose-3-O-sulfotransferase 2; galbeta1-3GalNAc 3'-sulfotransferase 2; Glycoprotein beta-Gal 3'-sulfotransferase 2
Gene Aliases: GAL3ST-2; GAL3ST2; GP3ST
UniProt ID: (Human) Q9H3Q3
Entrez Gene ID: (Human) 64090
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.