Invitrogen
Manufacturer Code:PA521580
Catalog # PIPA521580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 412 of GAL3ST1 (Uniprot IDNo.Q99999) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'-phosphoadenosine-5'-phosphosulfate:GalCer sulfotransferase; 3'-phosphoadenylylsulfate:galactosylceramide 3'-sulfotransferase; cerebroside (3'-phosphoadenylylsulfate:galactosylceramide 3') sulfotransferase; Cerebroside sulfotransferase; CST 3&prime-phosphoadenosine-5&prime-phosphosulfate:GalCer sulfotransferase 3&prime-phosphoadenylylsulfate:galactosylceramide 3&prime-sulfotransferase GalCer sulfotransferase cerebroside (3&prime-phosphoadenylylsulfate:galactosylceramide 3&prime) sulfotransferase cerebroside sulfotransferase galactosylceramide sulfotransferase; Galactosylceramide sulfotransferase; GalCer sulfotransferase
Gene Aliases: CST; GAL3ST1
UniProt ID: (Human) Q99999
Entrez Gene ID: (Human) 9514
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.