Abnova
Manufacturer Code:PAB1161
Catalog # 89-115-216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 1-300 <i>Saccharomyces cerevisiae</i> GAL11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae GAL11.
For Research Use Only
Protein Aliases: ABE1; Autonomous replication regulatory protein 3; Basal expression activator protein 1; Defective silencing suppressor protein 4; Gal11p; MED15; Mediator complex subunit 15; Mediator of RNA polymerase II transcription subunit 15; RAR3; SDS4; SPT13; Transcription regulatory protein GAL11; Ty insertion suppressor protein 13
Gene Aliases: ABE1; GAL11; MED15; RAR3; SDS4; SPT13; YOL051W
UniProt ID: (Baker's yeast) P19659
Entrez Gene ID: (Baker's yeast) 854106
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.