Abnova
Manufacturer Code:H00051083M01A
Catalog # 89-123-133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C1-G5 |
Immunogen | GAL (AAH30241 1 a.a. ~ 123 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant GAL.
For Research Use Only
Protein Aliases: Galanin; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin-message-associated peptide; galanin-related peptide; galanin/GMAP prepropeptide; GALN; GLNN; GMAP; MGC40167
Gene Aliases: ETL8; GAL; GAL-GMAP; GAL1; GALN; GLNN; GMAP
UniProt ID: (Human) P22466
Entrez Gene ID: (Human) 51083
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.