Invitrogen
Manufacturer Code:PA595500
Catalog # PIPA595500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human Galanin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Galanin; Galanin message-associated peptide; Galanin peptides; Galanin peptides Galanin Galanin message-associated peptide GMAP GAL GAL1 GALN GLNN; galanin prepropeptide; GMAP
Gene Aliases: Gal; Galn
UniProt ID: (Rat) P10683, (Mouse) P47212
Entrez Gene ID: (Rat) 29141, (Mouse) 14419
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.