Invitrogen
Manufacturer Code:PA519206
Catalog # PIPA519206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (HRSFSDKNGLTSK) corresponding to the internal amino acids of GAL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GAL galanin prepropeptide GALN GLNN GMAP MGC40167 galanin galanin-message-associated peptide galanin-related peptide; Galanin; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin-message-associated peptide; galanin-related peptide; galanin/GMAP prepropeptide; GMAP
Gene Aliases: ETL8; GAL; GAL-GMAP; GAL1; GALN; GLNN; GMAP
UniProt ID: (Human) P22466, (Rat) P10683, (Mouse) P47212
Entrez Gene ID: (Human) 51083, (Rat) 29141, (Mouse) 14419
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.