R&D Systems
Manufacturer Code:MAB6918
Catalog # MAB6918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 706306 |
Immunogen | <EM>E. coli</EM>-derived recombinant human GAK Phe258-Asp443 Accession # O14976 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: auxilin-2; Auxillin 2 cyclin G associated kinase cyclin-G-associated kinase EC 2.7.11.1 FLJ16629 FLJ40395 MGC99654; Cyclin-G-associated kinase
Gene Aliases: DNAJ26; DNAJC26; GAK
UniProt ID: (Human) O14976
Entrez Gene ID: (Human) 2580
Molecular Function: chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.