Novus Biologicals
Manufacturer Code:NBP232181
Catalog # NBP232181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 675 to 725 of human Cyclin G Associated Kinase using the numbering given in entry NP_005246.2 (GeneID 2580). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: auxilin-2; cyclin G associated kinase cyclin-G-associated kinase EC 2.7.11.1 FLJ16629 FLJ40395 MGC99654; Cyclin-G-associated kinase
Gene Aliases: DNAJ26; DNAJC26; GAK
UniProt ID: (Human) O14976
Entrez Gene ID: (Human) 2580
Molecular Function: chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.