Proteintech
Manufacturer Code:121471AP150UL
Catalog # 50-172-7747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GAK Fusion Protein Ag2795 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: auxilin-2; cyclin G associated kinase GAK; cyclin G-associated kinase; Cyclin-G-associated kinase
Gene Aliases: D130045N16Rik; DNAJ26; DNAJC26; GAK
UniProt ID: (Human) O14976, (Mouse) Q99KY4, (Rat) P97874
Entrez Gene ID: (Human) 2580, (Mouse) 231580, (Rat) 81659
Molecular Function: chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.