Proteintech
Manufacturer Code:125321AP150UL
Catalog # 50-172-8045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GAGE2D Fusion Protein Ag3230 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 4.8; Cancer/testis antigen 4.8 CT4.8 G antigen 2D G antigen 8 GAGE 8 GAGE2D GAGE8; CT4.8; G antigen 2D; G antigen 8; GAGE-2D; GAGE-8
Gene Aliases: CT4.8; GAGE-2D; GAGE-8; GAGE2D; GAGE8
UniProt ID: (Human) Q9UEU5
Entrez Gene ID: (Human) 729408
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.