Proteintech
Manufacturer Code:182401AP150UL
Catalog # 50-173-2066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GADL1 Fusion Protein Ag13005 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acidic amino acid decarboxylase GADL1; ADC; Aspartate 1-decarboxylase; CSADC; Cysteine sulfinic acid decarboxylase; GADL1 glutamate decarboxylase like 1; glutamate decarboxylase-like 1; Glutamate decarboxylase-like protein 1
Gene Aliases: 1110027M19Rik; ADC; CSADC; GADL1; HuADC; HuCSADC; RGD1565145
UniProt ID: (Human) Q6ZQY3, (Mouse) Q80WP8
Entrez Gene ID: (Human) 339896, (Rat) 367181, (Mouse) 73748
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.