Invitrogen
Manufacturer Code:PA5111409
Catalog # PIPA5111409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human GADD45GIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310040G17Rik 39S ribosomal protein L59 mitochondrial AI425883 CKBBP2 CKbetaBP2 CKII beta-associating protein CR6-interacting factor 1 CRIF1 GADD45GIP1 Growth arrest and DNA damage-inducible proteins-interacting protein 1 Mitochondrial large ribosomal subunit protein mL64 MRPL59 MRP-L59 p53-responsive gene 6 protein Papillomavirus L2-interacting nuclear protein 1 PLINP PLINP1 PLINP-1 PRG6; 39S ribosomal protein L59, mitochondrial; CKII beta binding protein 2; CKII beta-associating protein; CR6 interacting factor 1; CR6-interacting factor 1; CRIF1; Growth arrest and DNA damage-inducible proteins-interacting protein 1; growth arrest and DNA-damage-inducible, gamma interacting protein 1; growth arrest- and DNA damage-inducible GADD45G-interacting protein; Mitochondrial large ribosomal subunit protein mL64; MRP-L59; p53-responsive gene 6 protein; papillomavirus L2 interacting nuclear protein 1; Papillomavirus L2-interacting nuclear protein 1; PLINP
Gene Aliases: CKBBP2; CKbetaBP2; CRIF1; GADD45GIP1; MRP-L59; MRPL59; PLINP; PLINP-1; PLINP1; PRG6
UniProt ID: (Human) Q8TAE8
Entrez Gene ID: (Human) 90480
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.