Abnova
Manufacturer Code:H00090480K
Catalog # 89-203-915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human GADD45GIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human GADD45GIP1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 39S ribosomal protein L59, mitochondrial; CKBBP2 CRIF1 MGC4667 MGC4758 PLINP-1 PRG6 Plinp1; CKII beta binding protein 2; CKII beta-associating protein; CR6 interacting factor 1; CR6-interacting factor 1; CRIF1; Growth arrest and DNA damage-inducible proteins-interacting protein 1; growth arrest and DNA-damage-inducible, gamma interacting protein 1; growth arrest- and DNA damage-inducible GADD45G-interacting protein; Mitochondrial large ribosomal subunit protein mL64; MRP-L59; p53-responsive gene 6 protein; papillomavirus L2 interacting nuclear protein 1; Papillomavirus L2-interacting nuclear protein 1; PLINP
Gene Aliases: CKBBP2; CKbetaBP2; CRIF1; GADD45GIP1; MRP-L59; MRPL59; PLINP; PLINP-1; PLINP1; PRG6
UniProt ID: (Human) Q8TAE8
Entrez Gene ID: (Human) 90480
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.