Invitrogen
Manufacturer Code:PA565786
Catalog # PIPA565786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human GADD45A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDIT-1; DDIT1 GADD45; DNA damage-inducible transcript 1 protein; DNA damage-inducible transcript-1; DNA-damage-inducible transcript 1; Growth arrest and DNA damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible 45 alpha; growth arrest and DNA-damage-inducible, alpha
Gene Aliases: DDIT1; GADD45; GADD45A
UniProt ID: (Human) P24522
Entrez Gene ID: (Human) 1647
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.