Invitrogen
Manufacturer Code:PA5114736
Catalog # PIPA5114736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GADD45A corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA545191 DDIT 1 DDIT1 DDIT-1 DNA damage-inducible transcript 1 protein DNA damage-inducible transcript-1 DNA-damage inducible transcript 1 DNA-damage-inducible transcript 1 Gadd45 Gadd45a Gadd45alpha growth arrest and DNA damage inducible alpha growth arrest and DNA damage-inducible protein GADD45 alpha growth arrest and DNA-damage-inducible 45 alpha growth arrest and DNA-damage-inducible protein GADD45 alpha growth arrest and DNA-damage-inducible alpha; DDIT-1; DNA damage-inducible transcript 1 protein; DNA damage-inducible transcript-1; DNA-damage inducible transcript 1; DNA-damage-inducible transcript 1; Gadd45alpha; Growth arrest and DNA damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible 45 alpha; growth arrest and DNA-damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible, alpha
Gene Aliases: AA545191; DDIT1; GADD45; GADD45A
UniProt ID: (Human) P24522, (Mouse) P48316
Entrez Gene ID: (Human) 1647, (Mouse) 13197
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.