Novus Biologicals
Manufacturer Code:NBP271765H
Catalog # NBP271765H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D12 |
Immunogen | Full length recombinant protein of human GADD34 (NP_055145) produced in HEK293T cells. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GADD34Growth arrest and DNA damage-inducible protein GADD34 growth arrest and DNA-damage-inducible 34 Myeloid differentiation primary response protein MyD116 homolog protein phosphatase 1 regulatory subunit 15A protein phosphatase 1 regulatory (inhibitor) subunit 15A; Growth arrest and DNA damage-inducible protein GADD34; growth arrest and DNA-damage-inducible 34; Myeloid differentiation primary response protein MyD116 homolog; Protein phosphatase 1 regulatory subunit 15A; protein phosphatase 1, regulatory (inhibitor) subunit 15A; protein phosphatase 1, regulatory subunit 15A
Gene Aliases: GADD34; PPP1R15A
UniProt ID: (Human) O75807
Entrez Gene ID: (Human) 23645
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.