Invitrogen
Manufacturer Code:PA536054
Catalog # PIPA536054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 460-514 of human GAD-67 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 67 kDa glutamic acid decarboxylase; GAD-67; Glutamate decarboxylase 1; Glutamate decarboxylase 1 (brain); glutamate decarboxylase 1 (brain, 67kDa); Glutamate decarboxylase 1 67 kDa glutamic acid decarboxylase GAD 67 Glutamate decarboxylase 67 kDa isoform GAD1 GAD GAD67 GAD-1 GAD 1; glutamate decarboxylase 1 variant GAD67NT; Glutamate decarboxylase 67 kDa isoform; GLUTAMATE DECARBOXYLASE, 67 KD ISOFORM (GAD-67) (67 KD GLUTAMIC ACID DECARBOXYLASE); glutamic acid decarboxylase 1
Gene Aliases: CPSQ1; EP10; GAD; Gad-1; GAD1; GAD25; GAD44; GAD67; SCP
UniProt ID: (Human) Q99259, (Mouse) P48318, (Rat) P18088
Entrez Gene ID: (Human) 2571, (Mouse) 14415, (Rat) 24379
Molecular Function: decarboxylase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.