Invitrogen
Manufacturer Code:MA518053
Catalog # PIMA518053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 144 |
Immunogen | Synthetic peptide: GSGFWSFGSEDGSGDSEN conjugated to KLH corresponding to amino acids 4-22 of Human GAD65 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa glutamic acid decarboxylase; GAD-65; Glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); Glutamate decarboxylase 2 65 kDa glutamic acid decarboxylase Glutamate decarboxylase 65 kDa isoform 65 kDa glutamic acid decarboxylase; Glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase 2; glutamic acid decarboxylase 65
Gene Aliases: 6330404F12Rik; GAD(65); Gad-2; GAD2; GAD65
UniProt ID: (Human) Q05329, (Mouse) P48320, (Rat) Q05683
Entrez Gene ID: (Human) 2572, (Mouse) 14417, (Rat) 24380
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.