Invitrogen
Manufacturer Code:PA518369
Catalog # PIPA518369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TLEDNEERMSRLSK) corresponding to the internal amino acids of GAD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 65 kDa glutamic acid decarboxylase; GAD-65; GAD2 GAD65 glutamate decarboxylase 2 (pancreatic islets and brain 65kDa) Glutamate decarboxylase-2 (pancreas) glutamate decarboxylase 2 (pancreatic islets and brain 65kD) RP11-420F12.2 MGC161605 MGC161607 Glutamate decarboxylase-2 (pancreas) glutamate decarboxylase 2; Glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); Glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase 2; glutamic acid decarboxylase 65
Gene Aliases: 6330404F12Rik; GAD(65); Gad-2; GAD2; GAD65
UniProt ID: (Human) Q05329, (Mouse) P48320, (Rat) Q05683
Entrez Gene ID: (Human) 2572, (Mouse) 14417, (Rat) 24380
Molecular Function: decarboxylase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.