Novus Biologicals
Manufacturer Code:NBP148272
Catalog # NBP148272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G9 |
Immunogen | Full length human recombinant protein of human GAD1 (NP_000808) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 67 kDa glutamic acid decarboxylase; 67kD) EC 4.1.1 FLJ45882 GAD25 glutamate decarboxylase 1 (brain 67kDa) Glutamate decarboxylase 67 kDa isoform; GAD-67; Glutamate decarboxylase 1; glutamate decarboxylase 1 (brain, 67kDa); Glutamate decarboxylase 67 kDa isoform
Gene Aliases: CPSQ1; GAD; GAD1; GAD67; SCP
UniProt ID: (Human) Q99259
Entrez Gene ID: (Human) 2571
Molecular Function: decarboxylase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.