Proteintech
Manufacturer Code:199581AP150UL
Catalog # 50-553-514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAD20 GAC GAC-specific GLS GLS1 glutaminase C K glutaminase KIAA0838 L glutamine amidohydrolase; GLS; glutaminase C; Glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; K-glutaminase; L-glutamine amidohydrolase
Gene Aliases: 6330442B14; AAD20; AI314027; B230365M23Rik; GAC; GAM; GLS; GLS1; Glut; KGA; KIAA0838; RATGLUT
UniProt ID: (Human) O94925, (Mouse) D3Z7P3, (Rat) P13264
Entrez Gene ID: (Human) 2744, (Mouse) 14660, (Rat) 24398
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.