Invitrogen
Manufacturer Code:PA546830
Catalog # PIPA546830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GABRP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit pi; GABA(A) receptor, pi; GABA-A receptor pi subunit; Gamma-aminobutyric acid (GABA) A receptor pi MGC126386 MGC126387; gamma-aminobutyric acid (GABA) A receptor, pi; gamma-aminobutyric acid (GABA-A) receptor, pi; gamma-aminobutyric acid A receptor, pi; Gamma-aminobutyric acid receptor subunit pi; gamma-aminobutyric acid type A receptor pi subunit
Gene Aliases: GABRP
UniProt ID: (Human) O00591, (Mouse) Q8QZW7, (Rat) O09028
Entrez Gene ID: (Human) 2568, (Horse) 100070350, (Rabbit) 100339970, (Dog) 489124, (Mouse) 216643, (Rat) 81658
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.