Invitrogen
Manufacturer Code:PA595653
Catalog # PIPA595653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fusion protein from the cytoplasmic loop of the gamma 2 subunit |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit gamma-2; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 2; gamma-aminobutyric acid A receptor, gamma 2; Gamma-aminobutyric acid receptor subunit gamma-2
Gene Aliases: GABAA-R; Gabrg-2; Gabrg2; gamma2
UniProt ID: (Mouse) P22723, (Rat) P18508
Entrez Gene ID: (Mouse) 14406, (Rat) 29709
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.