Invitrogen
Manufacturer Code:PA541086
Catalog # PIPA541086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human GABRG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; gamma-aminobutyric acid A receptor, gamma 2; Gamma-aminobutyric acid (GABA) A receptor gamma 2 CAE2 ECA2 GEFSP3; gamma-aminobutyric acid (GABA) A receptor, gamma 2; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 2; gamma-aminobutyric acid A receptor, gamma 2; Gamma-aminobutyric acid receptor subunit gamma-2; gamma-aminobutyric acid type A receptor gamma2 subunit
Gene Aliases: CAE2; ECA2; GABAA-R; Gabrg-2; GABRG2; gamma2; GEFSP3; si:ch211-145n14.1
UniProt ID: (Human) P18507, (Bovine) P22300, (Mouse) P22723, (Rat) P18508
Entrez Gene ID: (Human) 2566, (Horse) 100059571, (Rabbit) 100357302, (Dog) 489141, (Bovine) 282240, (Guinea pig) 100723005, (Mouse) 14406, (Rat) 29709, (Zebrafish) 553402
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.