Invitrogen
Manufacturer Code:PA523698
Catalog # PIPA523698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 198-224 amino acids from the central region of human GABRG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAE2 ECA2 GEFSP3 GABA(A) receptor subunit gamma-2 GABA(A) receptor gamma 2 gamma-aminobutyric acid receptor subunit gamma-2; GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; Gamma-aminobutyric acid receptor subunit gamma-2
Gene Aliases: CAE2; ECA2; GABAA-R; Gabrg-2; GABRG2; gamma2; GEFSP3
UniProt ID: (Human) P18507, (Mouse) P22723
Entrez Gene ID: (Human) 2566, (Mouse) 14406
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.